4-Chinolinol
Lieferant: Molekula
|
Achtung
|
Synonyme:
Quinolin-4-ol, 4-Hydroxyquinoline
76234561-5G
MOLE76234561-5GEA
0
EUR
MOLE76234561-5G
4-Chinolinol
4-Chinolinol
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 202…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006777 CAS-Nummer: 611-36-9 EINECS: 210-268-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...