(S)-(+)-1,1'-Binaphthyl-2,2'-diylhydrogenphosphat

Lieferant: Molekula

Synonyme: (S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

76943966-1G
MOLE76943966-1GEA 0 EUR
MOLE76943966-1G
(S)-(+)-1,1'-Binaphthyl-2,2'-diylhydrogenphosphat
(S)-(+)-1,1'-Binaphthyl-2,2'-diylhydrogenphosphat
Formel: C₂₀H₁₃O₄P
Molecular Weight: 348,289 g/mol
MDL: MFCD00010045
CAS-Nummer: 35193-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR