7-(2,3-Dihydroxypropyl)theophyllin

Lieferant: Molekula

Synonyme: Dyphylline, 7-(2,3-Dihydroxypropyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione

77994840-25MG
MOLE77994840-25MGEA 0 EUR
MOLE77994840-25MG
7-(2,3-Dihydroxypropyl)theophyllin
7-(2,3-Dihydroxypropyl)theophyllin
Formel: C₁₀H₁₄N₄O₄
Molecular Weight: 254,24 g/mol
MDL: MFCD00005759
CAS-Nummer: 479-18-5
EINECS: 207-526-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR