8-Hydroxy-5-chinolinsulfonsäure

Lieferant: Molekula

78011644-500G
MOLE78011644-500GEA 0 EUR
MOLE78011644-500G
8-Hydroxy-5-chinolinsulfonsäure
8-Hydroxyquinoline-5-sulphonic acid
Formel: C₉H₇NO₄S
CAS-Nummer: 84-88-8
EINECS: 201-570-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR