Guanosin-5'-diphosphat Dinatriumsalz

Lieferant: Molekula

Synonyme: ((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate, disodium salt, Guanosine 5'-diphosphate disodium salt

78355753-100MG
MOLE78355753-100MGEA 125 EUR
MOLE78355753-100MG
Guanosin-5'-diphosphat Dinatriumsalz
Guanosin-5'-diphosphat Dinatriumsalz
Formel: C₁₀H₁₃N₅Na₂O₁₁P₂
Molecular Weight: 487,17 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00070121
CAS-Nummer: 7415-69-2
EINECS: 231-026-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR