Triethylbutylammoniumbromid
Lieferant: Molekula
Synonyme:
(1-Butyl)triethylammonium bromide
80265642-100G
MOLE80265642-100GEA
0
EUR
MOLE80265642-100G
Triethylbutylammoniumbromid
Triethylbutylammoniumbromid
Formel:
C₁₀H₂₄BrN Molecular Weight: 238,21 g/mol |
MDL:
MFCD00055541 CAS-Nummer: 13028-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...