(R)-(-)-2-Chlormandelsäure

Lieferant: Molekula

Synonyme: (R)-2-(2-Chlorophenyl)-2-hydroxyacetic acid

81595483-500MG
MOLE81595483-500MGEA 0 EUR
MOLE81595483-500MG
(R)-(-)-2-Chlormandelsäure
(R)-(-)-2-Chlormandelsäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD00798429
CAS-Nummer: 52950-18-2
EINECS: 444-040-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR