Acridingelb

Lieferant: Molekula

Synonyme: 3,6-Diamino-2,7-dimethylacridine monohydrochloride, CI 46025, Acridine yellow, Acridine Yellow G

82602455-25G
MOLE82602455-25GEA 163 EUR
MOLE82602455-25G
Acridingelb
Acridingelb G
Formel: C₁₅H₁₅N₃·HCl
Molecular Weight: 273,76 g/mol
MDL: MFCD00012661
CAS-Nummer: 135-49-9
EINECS: 205-194-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR