Methyl-3,5-dimethoxybenzoat

Lieferant: Molekula

Synonyme: 3,5-Dimethoxybenzoesäuremethylester

82959542-100G
MOLE82959542-100GEA 0 EUR
MOLE82959542-100G
Methyl-3,5-dimethoxybenzoat
Methyl-3,5-dimethoxybenzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00008432
CAS-Nummer: 2150-37-0
EINECS: 218-423-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR