(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure

Lieferant: Molekula

Synonyme: Tarenflurbil, (R)-2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, (R)-(-)-Flurbiprofen

83025196-500MG
MOLE83025196-500MGEA 0 EUR
MOLE83025196-500MG
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244,260 g/mol
MDL: MFCD00869714
CAS-Nummer: 51543-40-9
EINECS: 257-264-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR