(2S)-2-Pyrrolidinylmethanol

Lieferant: Molekula

Synonyme: (S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol

83380641-5G
MOLE83380641-5GEA 110 EUR
MOLE83380641-5G
(2S)-2-Pyrrolidinylmethanol
(S)-(+)-Prolinol
Formel: C₅H₁₁NO
Molecular Weight: 101,15 g/mol
MDL: MFCD00005255
CAS-Nummer: 23356-96-9
EINECS: 245-605-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR