Tetrabutylammoniumtetrafluoroborat
Lieferant: Molekula
|
Achtung
|
Synonyme:
Tetrabutylammoniumtetrafluoroborate
84066585-5G
MOLE84066585-5GEA
0
EUR
MOLE84066585-5G
Tetrabutylammoniumtetrafluoroborat
Tetrabutylammoniumtetrafluoroborat
Formel:
C₁₆H₃₆BF₄N Molecular Weight: 329,27 g/mol Schmelzpunkt: 159…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011634 CAS-Nummer: 429-42-5 EINECS: 207-058-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...