7-Indolcarbaldehyd

Lieferant: Molekula

Synonyme: Indol-7-carboxaldehyd

84400316-1G
MOLE84400316-1GEA 0 EUR
MOLE84400316-1G
7-Indolcarbaldehyd
7-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD01318152
CAS-Nummer: 1074-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR