2',3'-Didesoxycytidin
Lieferant: Molekula
Synonyme:
4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine
84941822-100MG
MOLE84941822-100MGEA
353
EUR
MOLE84941822-100MG
2',3'-Didesoxycytidin
2',3'-Didesoxycytidin
Formel:
C₉H₁₃N₃O₃ Molecular Weight: 211,22 g/mol |
MDL:
MFCD00012188 CAS-Nummer: 7481-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...