Adenin Hydrochlorid

Lieferant: Molekula
Achtung

Synonyme: Purin-6-amin Hydrochlorid, 1H-Adenin Hydrochlorid, 9H-Purin-6-amin Hydrochlorid, 6-Aminopurin Hydrochlorid

85430679-5G
MOLE85430679-5GEA 71.6 EUR
MOLE85430679-5G
Adenin Hydrochlorid
Adenin Hydrochlorid
Formel: C₅H₅N₅·HCl
Molecular Weight: 171,59 g/mol
MDL: MFCD00150864
CAS-Nummer: 2922-28-3
EINECS: 220-867-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR