4-Hydroxyzimtsäure

Lieferant: Molekula
Achtung

Synonyme: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid

86815064-100G
MOLE86815064-100GEA 229 EUR
MOLE86815064-100G
4-Hydroxyzimtsäure
4-Hydroxyzimtsäure
Formel: C₉H₈O₃
Molecular Weight: 164,16 g/mol
MDL: MFCD00004399
CAS-Nummer: 7400-08-0
EINECS: 231-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR