1,3-Phenyldiboronsäure

Lieferant: Molekula

Synonyme: 1,3-Benzoldiboronsäure, Benzol-1,3-diboronsäure, m-Phenylenbisboronsäure

86868254-1G
MOLE86868254-1GEA 0 EUR
MOLE86868254-1G
1,3-Phenyldiboronsäure
1,3-Phenyldiboronsäure
Formel: C₆H₈B₂O₄
Molecular Weight: 165,75 g/mol
MDL: MFCD03092915
CAS-Nummer: 4612-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR