6-Chlornicotinonitril

Lieferant: Molekula

Synonyme: 2-Chlor-5-pyridincarbonitril, 6-Chlornicotinsäurenitril, 2-Chlor-5-cyanpyridin, 2-Chlor-5-pyridincarbonsäurenitril

87085479-5G
MOLE87085479-5GEA 0 EUR
MOLE87085479-5G
6-Chlornicotinonitril
6-Chlornicotinonitril
Formel: C₆H₃ClN₂
MDL: MFCD00084941
CAS-Nummer: 33252-28-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR