4'-Nitrobenzo-18-krone-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecan)

Lieferant: Molekula

Synonyme: 18-Nitro-2,3,5,6,8,9,11,12,14,15-decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine

88364334-1G
MOLE88364334-1GEA 0 EUR
MOLE88364334-1G
4'-Nitrobenzo-18-krone-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecan)
4'-Nitrobenzo-18-krone-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Formel: C₁₆H₂₃NO₈
Molecular Weight: 357,35 g/mol
MDL: MFCD00143207
CAS-Nummer: 53408-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR