Pluronic® F-68

Lieferant: Molekula

Synonyme: PEG-PPG-PEG, Kolliphor® P 188, Synperonic® F68, Lutrol® F68, Pluronic® F-68

89483155-500G
MOLE89483155-500GEA 209 EUR
MOLE89483155-500G
Pluronic® F-68
Kolliphor® P 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)
Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Molecular Weight: 8400
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR