2-Aminoethyldihydrogenphosphat

Lieferant: Molekula

Synonyme: O-Phosphorylethanolamine

89847060-25G
MOLE89847060-25GEA 0 EUR
MOLE89847060-25G
2-Aminoethyldihydrogenphosphat
2-Aminoethyldihydrogenphosphat
Formel: C₂H₈NO₄P
Molecular Weight: 141.0630 g/mol
MDL: MFCD00008178
CAS-Nummer: 1071-23-4
EINECS: 213-988-5
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR