5-(Benzylthio)-1H-tetrazol

Lieferant: Molekula

Synonyme: BMT, BTT

89914250-25G
MOLE89914250-25GEA 174 EUR
MOLE89914250-25G
5-(Benzylthio)-1H-tetrazol
5-(Benzylthio)-1H-tetrazol
Formel: C₈H₈N₄S
Molecular Weight: 192.24 g/mol
MDL: MFCD00068731
CAS-Nummer: 21871-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR