Isochinolin-4-amin

Lieferant: Molekula

Synonyme: 4-Aminoisoquinoline, Isoquinolin-4-amine

89985050
MOLE89985050EA 119 EUR
MOLE89985050
Isochinolin-4-amin
Isochinolin-4-amin
Formel: C₉H₈N₂
Molecular Weight: 144.1700 g/mol
MDL: MFCD00034752
CAS-Nummer: 23687-25-4
EINECS: 245-823-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR