n-Octyl-β-D-maltosid

Lieferant: Molekula

90033684
MOLE90033684EA 0 EUR
MOLE90033684
n-Octyl-β-D-maltosid
n-Octyl-β-D-maltosid
Formel: C₂₀H₃₈O₁₁
CAS-Nummer: 82494-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR