(S)-(+)-2-Hexanol
Lieferant: Molekula
Synonyme:
(S)-(+)-2-Hydroxyhexan, (S)-(+)-Butyl-methylcarbinol, (S)-(+)-Methyl-butylcarbinol
90035495-1G
MOLE90035495-1GEA
0
EUR
MOLE90035495-1G
(S)-(+)-2-Hexanol
(S)-(+)-2-Hexanol
Formel:
C₆H₁₄O Siedepunkt: 137 °C (1013 hPa) Dichte: 0,81 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065955 CAS-Nummer: 52019-78-0 UN: 2282 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...