2-Hydroxy-3-nitropyridin
Lieferant: Molekula
Synonyme:
3-Nitro-2-pyridinol
90035566
MOLE90035566EA
0
EUR
MOLE90035566
2-Hydroxy-3-nitropyridin
2-Hydroxy-3-nitropyridine
Formel:
C₅H₄N₂O₃ Molecular Weight: 140,1 g/mol Schmelzpunkt: 224 °C |
MDL:
MFCD00006270 CAS-Nummer: 6332-56-5 EINECS: 228-709-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...