Scandium(III)nitrat-Hydrat
Lieferant: Molekula
Synonyme:
Scandiumtrinitrat
C76519134
MOLEC76519134EA
0
EUR
MOLEC76519134
Scandium(III)nitrat-Hydrat
Scandium(III)nitrat-Hydrat
Formel:
Sc(NO₃)₃·xH₂O Molecular Weight: 230,97 g/mol (anhy) |
MDL:
MFCD00011220 CAS-Nummer: 107552-14-7 EINECS: 236-701-5 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...