TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)

Lieferant: Molekula

Synonyme: TBTU

M10255546
MOLEM10255546EA 0 EUR
MOLEM10255546
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
Formel: C₁₁H₁₆BF₄N₅O
Molecular Weight: 321,1 g/mol
MDL: MFCD00077413
CAS-Nummer: 125700-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR