EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)

Lieferant: Molekula

M10325546
MOLEM10325546EA 0 EUR
MOLEM10325546
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
Formel: C₁₄H₁₇NO₃
MDL: MFCD00006703
CAS-Nummer: 16357-59-8
EINECS: 240-418-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR