1-Amino-4-naphthol Hydrochlorid
Lieferant: Molekula
Synonyme:
4-Amino-1-naphthol hydrochloride
M10434262
MOLEM10434262EA
0
EUR
MOLEM10434262
1-Amino-4-naphthol Hydrochlorid
1-Amino-4-naphthol Hydrochlorid
Formel:
C₁₀H₁₀ClNO Molecular Weight: 195,65 g/mol |
MDL:
MFCD00012562 CAS-Nummer: 5959-56-8 EINECS: 227-727-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...