HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)

Lieferant: Molekula

Synonyme: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

M10466417
MOLEM10466417EA 0 EUR
MOLEM10466417
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
Formel: C₈H₁₈N₂O₄S
Molecular Weight: 238.31 g/mol
Schmelzpunkt: 210…215 °C
Dichte: 1.448 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006158
CAS-Nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR