TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)

Lieferant: Molekula

Synonyme: Tris(hydroxymethyl)methylamin Hydrochlorid, 2-Amino-2-(hydroxymethyl)propan-1,3-diolhydrochlorid, 2-Amino-2-(hydroxymethyl)-1,3-propandiol HCl, Tris(hydroxymethyl)methylammoniumchlorid, TRIS HCl, THAM HCl, Trometamol Hydrochlorid

M10696487
MOLEM10696487EA 0 EUR
MOLEM10696487
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
Formel: NH₂C(CH₂OH)₃·HCl
Molecular Weight: 157.6 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012590
CAS-Nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR