X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)

Lieferant: Molekula

M10742126
MOLEM10742126EA 0 EUR
MOLEM10742126
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel: C₁₄H₁₅BrClNO₆
Schmelzpunkt: 237…239 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005666
CAS-Nummer: 7240-90-6
EINECS: 230-640-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR