TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
Lieferant: Molekula
|
Achtung
|
Synonyme:
TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid
M10813371
MOLEM10813371EA
0
EUR
MOLEM10813371
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
Formel:
C₆H₁₅NO₆S Molecular Weight: 229,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007532 CAS-Nummer: 7365-44-8 EINECS: 230-906-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...