4,4'-Dimethoxytritylchlorid
Lieferant: Molekula
Synonyme:
DMT-Cl, 4,4'-Dimethoxytriphenylmethyl chloride
M10955409
MOLEM10955409EA
0
EUR
MOLEM10955409
4,4'-Dimethoxytritylchlorid
4,4'-Dimethoxytritylchlorid
Formel:
C₂₁H₁₉ClO₂ Molecular Weight: 338,8 g/mol |
MDL:
MFCD00008409 CAS-Nummer: 40615-36-9 EINECS: 255-002-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...