(R)-(-)-1-Methoxy-2-propanol

Lieferant: Molekula

Synonyme: (R)-(-)-Propylenglycolmonomethylether

M10995842
MOLEM10995842EA 0 EUR
MOLEM10995842
(R)-(-)-1-Methoxy-2-propanol
(R)-(-)-1-Methoxy-2-propanol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632587
CAS-Nummer: 4984-22-9
UN: 3092
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR