Guanidiniumsulfat

Lieferant: Molekula

Synonyme: Guanidine sulphate

M11055102
MOLEM11055102EA 0 EUR
MOLEM11055102
Guanidiniumsulfat
Guanidinium sulphate
Formel: C₂H₁₂N₆O₄S
Molecular Weight: 216,22 g/mol
MDL: MFCD00013115
CAS-Nummer: 594-14-9
EINECS: 209-826-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR