9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
Lieferant: Molekula
Synonyme:
(9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl
M11086183
MOLEM11086183EA
0
EUR
MOLEM11086183
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
9-Fluorenylmethylchlorformiat (Fmoc-Chlorid)
Formel:
C₁₅H₁₁ClO₂ Molecular Weight: 258.7 g/mol |
MDL:
MFCD00001138 CAS-Nummer: 28920-43-6 EINECS: 249-313-6 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...