Oxalsäuremethylesterchlorid
Lieferant: Molekula
Synonyme:
Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate
M11177203
MOLEM11177203EA
0
EUR
MOLEM11177203
Oxalsäuremethylesterchlorid
Oxalsäuremethylesterchlorid
Formel:
C₃H₃ClO₃ Molecular Weight: 122,51 g/mol |
MDL:
MFCD00000705 CAS-Nummer: 5781-53-3 EINECS: 227-307-4 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...