Dimidiumbromid

Lieferant: Molekula
Achtung

Synonyme: 3,8-Diamino-5-methyl-6-phenylphenanthridiniumbromid, 2,7-Diamino-10-methyl-9-phenylphenanthridiniumbromid

M11783520
MOLEM11783520EA 0 EUR
MOLEM11783520
Dimidiumbromid
Dimidiumbromid
Formel: C₂₀H₁₈BrN₃
Molecular Weight: 380,29 g/mol
Schmelzpunkt: 243…248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011757
CAS-Nummer: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR