Cycloheximid

Lieferant: Molekula
Gefahr

M11784988
MOLEM11784988EA 0 EUR
MOLEM11784988
Cycloheximid
Cycloheximid
Formel: C₁₅H₂₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00082346
CAS-Nummer: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR