2-Amino-5-fluorpyridin

Lieferant: Molekula

M11804620
MOLEM11804620EA 0 EUR
MOLEM11804620
2-Amino-5-fluorpyridin
2-Amino-5-fluorpyridin
Formel: C₅H₅FN₂
MDL: MFCD01861120
CAS-Nummer: 21717-96-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR