Bicin

Lieferant: Molekula

Synonyme: N,N-Bis(2-hydroxyethyl)glycin

M12063561
MOLEM12063561EA 0 EUR
MOLEM12063561
Bicin
Bicin
Formel: C₆H₁₃NO₄
Molecular Weight: 163,17 g/mol
Schmelzpunkt: 185…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004295
CAS-Nummer: 150-25-4
EINECS: 205-755-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR