Methyl-3-indolcarboxylat

Lieferant: Molekula

M12121508
MOLEM12121508EA 0 EUR
MOLEM12121508
Methyl-3-indolcarboxylat
Methyl-3-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00189407
CAS-Nummer: 942-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR