L-Methionin
Lieferant: Molekula
Synonyme:
H-Met-OH, (S)-(-)-2-Amino-4-(methylmercapto)buttersäure, (S)-(-)-2-Amino-4-(methylthio)buttersäure, L(-)-Methionin, (S)-(-)-Methionin
M12412187
MOLEM12412187EA
0
EUR
MOLEM12412187
L-Methionin
L-Methionin
Formel:
C₅H₁₁NO₂S Molecular Weight: 149.21 g/mol Siedepunkt: 278 Schmelzpunkt: 136 Dichte: 1.373 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063097 CAS-Nummer: 63-68-3 EINECS: 200-562-9 Merck Index: 13,06004 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...