CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Lieferant: Molekula
Synonyme:
3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid
M12546873
MOLEM12546873EA
0
EUR
MOLEM12546873
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel:
C₉H₁₉NO₄S Molecular Weight: 237,32 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041778 CAS-Nummer: 73463-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...