2,7-Dinitro-9-fluorenon

Lieferant: Molekula

M12944437
MOLEM12944437EA 0 EUR
MOLEM12944437
2,7-Dinitro-9-fluorenon
2,7-Dinitro-9-fluorenon
Formel: C₁₃H₆N₂O₅
Molecular Weight: 270,2 g/mol
Schmelzpunkt: 296 °C
MDL: MFCD00001153
CAS-Nummer: 31551-45-8
EINECS: 250-695-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR