5-Methoxytryptamin

Lieferant: Molekula

M12986820
MOLEM12986820EA 0 EUR
MOLEM12986820
5-Methoxytryptamin
5-Methoxytryptamin
Formel: C₁₁H₁₄N₂O
Molecular Weight: 190,24 g/mol
MDL: MFCD00005662
CAS-Nummer: 608-07-1
EINECS: 210-153-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR