DIPSO

Lieferant: Molekula

Synonyme: 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid

M12996752
MOLEM12996752EA 0 EUR
MOLEM12996752
DIPSO
DIPSO
Formel: C₇H₁₇NO₆S
Molecular Weight: 261.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038353
CAS-Nummer: 68399-80-4
EINECS: 269-992-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR