MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)

Lieferant: Molekula
Achtung

Synonyme: 2-Hydroxy-3-morpholinopropane-1-sulphonic acid

M13616262
MOLEM13616262EA 0 EUR
MOLEM13616262
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)
Formel: C₇H₁₅NO₅S
Molecular Weight: 225,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009629
CAS-Nummer: 68399-77-9
EINECS: 269-989-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR